2-Mercapto-4-phenylthiazol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD76224-1GEA 25 EUR
BLDPBD76224-1G BLDPBD76224-25G BLDPBD76224-5G
2-Mercapto-4-phenylthiazol 98%
2-Mercapto-4-phenylthiazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇NS₂
Molecular Weight: 193,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014508
CAS-Nummer: 2103-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR