2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan 97%
|
|
BLDPBD76104-5GEA
7
EUR
BLDPBD76104-5G
BLDPBD76104-1G
BLDPBD76104-25G
BLDPBD76104-10G
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan 97%
2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₉H₁₉BO₃ Molecular Weight: 186,06 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00192241 CAS-Nummer: 61676-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...