2-Iod-3-nitrotoluol 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD75960-1GEA 13 EUR
BLDPBD75960-1G BLDPBD75960-25G BLDPBD75960-100G BLDPBD75960-5G BLDPBD75960-10G
2-Iod-3-nitrotoluol 97%
2-Iod-3-nitrotoluol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₆INO₂
Molecular Weight: 263,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00091040
CAS-Nummer: 6277-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR