4',5'-Methylendioxy-2'-nitroacetophenon 98+%

Lieferant: BLD PHARMATECH GMBH

BLDPBD7492-5GEA 120 EUR
BLDPBD7492-5G BLDPBD7492-250MG BLDPBD7492-1G
4',5'-Methylendioxy-2'-nitroacetophenon 98+%
4',5'-Methylendioxy-2'-nitroacetophenon

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₇NO₅
Molecular Weight: 209,16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00053045
CAS-Nummer: 56136-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR