4-Amino-3-fluorphenol Hydrochlorid 96%

BLDPBD74860-25GEA 136 EUR
BLDPBD74860-25G BLDPBD74860-10G BLDPBD74860-5G BLDPBD74860-1G
4-Amino-3-fluorphenol Hydrochlorid 96%
4-Amino-3-fluorphenol Hydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₇ClFNO
Molecular Weight: 163,5773 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632189
CAS-Nummer: 18266-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR