2-Amino-4-(4-chlorphenyl)thiazol 98%

BLDPBD7386-5GEA 45 EUR
BLDPBD7386-5G BLDPBD7386-10G BLDPBD7386-1G BLDPBD7386-250MG BLDPBD7386-25G
2-Amino-4-(4-chlorphenyl)thiazol 98%
2-Amino-4-(4-chlorophenyl)thiazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₇ClN₂S
Molecular Weight: 210,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047058
CAS-Nummer: 2103-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR