2-Chlorethyl-phenylsulfid 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD73602-100GEA 267 EUR
BLDPBD73602-100G BLDPBD73602-5G BLDPBD73602-1G BLDPBD73602-25G BLDPBD73602-10G
2-Chlorethyl-phenylsulfid 98%
2-Chlorethyl-phenylsulfid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₉ClS
Molecular Weight: 172,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013691
CAS-Nummer: 5535-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR