2-Chlor-6-methoxynicotinsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD73400-5GEA 429 EUR
BLDPBD73400-5G BLDPBD73400-250MG BLDPBD73400-100MG BLDPBD73400-1G BLDPBD73400-25G
2-Chlor-6-methoxynicotinsäure 95%
2-Chlor-6-methoxynicotinsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₆NO₃Cl
Molecular Weight: 187,5804 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08236845
CAS-Nummer: 503000-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR