2'-Chlor-6'-fluor-3'-methylacetophenon 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD73332-250MGEA 7 EUR
BLDPBD73332-250MG
2'-Chlor-6'-fluor-3'-methylacetophenon 98%
2'-Chlor-6'-fluor-3'-methylacetophenon

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₈ClFO
Molecular Weight: 186,61 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01631368
CAS-Nummer: 261762-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR