2-Chinoxalincarbonsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD7316-100GEA 178 EUR
BLDPBD7316-100G BLDPBD7316-25G BLDPBD7316-5G BLDPBD7316-10G BLDPBD7316-500G BLDPBD7316-1G
2-Chinoxalincarbonsäure 97%
2-Quinoxalinecarboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₆N₂O₂
Molecular Weight: 174,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012334
CAS-Nummer: 879-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR