2-Brom-4-iodtoluol 98%

BLDPBD71814-5GEA 18 EUR
BLDPBD71814-5G BLDPBD71814-10G BLDPBD71814-25G BLDPBD71814-100G BLDPBD71814-1G
2-Brom-4-iodtoluol 98%
2-Brom-4-iodtoluol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₆BrI
Molecular Weight: 296,9300 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07779014
CAS-Nummer: 26670-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR