2-Amino-4'-fluorbenzophenon 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD70168-25GEA 24 EUR
BLDPBD70168-25G BLDPBD70168-5G BLDPBD70168-100G BLDPBD70168-10G
2-Amino-4'-fluorbenzophenon 95%
2-Amino-4'-fluorbenzophenon

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₃H₁₀FNO
Molecular Weight: 215,2230 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658166
CAS-Nummer: 3800-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR