2-Isobutyl-3-methoxypyrazin 97%

BLDPBD6998-5GEA 38 EUR
BLDPBD6998-5G
2-Isobutyl-3-methoxypyrazin 97%
2-Isobutyl-3-methoxypyrazin

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₄N₂O
Molecular Weight: 166,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006128
CAS-Nummer: 24683-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR