4-Isopropoxyphenylboronsäure 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD6939-25GEA 42 EUR
BLDPBD6939-25G BLDPBD6939-5G BLDPBD6939-10G BLDPBD6939-0.025G BLDPBD6939-100G
4-Isopropoxyphenylboronsäure 97%
4-Isopropoxyphenylboronsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₁₃BO₃
Molecular Weight: 180,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03427051
CAS-Nummer: 153624-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR