4-Isopropylphenylboronsäure 95%

BLDPBD6933-10GEA 38 EUR
BLDPBD6933-10G BLDPBD6933-5G BLDPBD6933-100G BLDPBD6933-25G BLDPBD6933-1G
4-Isopropylphenylboronsäure 95%
4-Isopropylphenylboronsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₁₃BO₂
Molecular Weight: 164,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01074614
CAS-Nummer: 16152-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR