2-Isopropylphenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD6931-5GEA 53 EUR
BLDPBD6931-5G BLDPBD6931-250MG BLDPBD6931-25G BLDPBD6931-100G BLDPBD6931-1G BLDPBD6931-10G
2-Isopropylphenylboronsäure 98%
2-Isopropylphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₃BO₂
Molecular Weight: 164,01 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03411937
CAS-Nummer: 89787-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR