2,8-Dimethylimidazo[1,2-a]pyridin-3-carbonsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD68692-100MGEA 43 EUR
BLDPBD68692-100MG BLDPBD68692-1G BLDPBD68692-250MG
2,8-Dimethylimidazo[1,2-a]pyridin-3-carbonsäure 95%
2,8-Dimethylimidazo[1,2-a]pyridin-3-carbonsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269632
CAS-Nummer: 874605-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR