2,6-Dichlorpyridin-N-oxid 98%
|
|
BLDPBD68230-25GEA
224
EUR
BLDPBD68230-25G
BLDPBD68230-100G
BLDPBD68230-1G
BLDPBD68230-5G
2,6-Dichlorpyridin-N-oxid 98%
2,6-Dichlorpyridin-N-oxid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₅H₃Cl₂NO Molecular Weight: 163,99 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00130246 CAS-Nummer: 2587-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...