1-Methyl-2-indolboronsäure 98%

BLDPBD6578-25GEA 465 EUR
BLDPBD6578-0.025G BLDPBD6578-25G BLDPBD6578-5G
1-Methyl-2-indolboronsäure 98%
1-Methyl-2-indoleboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₉H₁₀BNO₂
Molecular Weight: 174,99 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01114668
CAS-Nummer: 191162-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR