3-(Ethoxycarbonyl)-4-fluorphenylboronsäure 97%

BLDPBD6576-1GEA 16 EUR
BLDPBD6576-1G BLDPBD6576-250MG BLDPBD6576-5G BLDPBD6576-10G
3-(Ethoxycarbonyl)-4-fluorphenylboronsäure 97%
3-(Ethoxycarbonyl)-4-fluorphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₀BFO₄
Molecular Weight: 211,9827 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08061836
CAS-Nummer: 874219-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR