2,3,6-Trimethylchinoxalin 95%
Lieferant: BLD PHARMATECH GMBH
|
|
BLDPBD65268-1GEA
13
EUR
BLDPBD65268-1G
BLDPBD65268-5G
BLDPBD65268-25G
BLDPBD65268-100G
2,3,6-Trimethylchinoxalin 95%
2,3,6-Trimethylquinoxaline
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₁H₁₂N₂ Molecular Weight: 172,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047593 CAS-Nummer: 17635-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...