2,3,6-Trichloropyridine 96%

BLDPBD65236-500GEA 526 EUR
BLDPBD65236-500G BLDPBD65236-10G BLDPBD65236-25G BLDPBD65236-100G BLDPBD65236-5G
2,3,6-Trichloropyridine 96%
2,3,6-Trichloropyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₅H₂Cl₃N
Molecular Weight: 182,43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463516
CAS-Nummer: 6515-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR