2-Formyl-5-methoxyphenylboronsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD6453-5GEA 201 EUR
BLDPBD6453-5G BLDPBD6453-1G BLDPBD6453-250MG BLDPBD6453-100MG
2-Formyl-5-methoxyphenylboronsäure 98%
2-Formyl-5-methoxyphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₉BO₄
Molecular Weight: 179,9657 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03001335
CAS-Nummer: 40138-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR