2,2,2-Trifluorethylbutyrat 98%
|
|
BLDPBD63880-5GEA
32
EUR
BLDPBD63880-5G
BLDPBD63880-25G
BLDPBD63880-1G
2,2,2-Trifluorethylbutyrat 98%
2,2,2-Trifluorethylbutyrat
Storage condition: Sealed in dry, Room Temperature
Formel:
C₆H₉F₃O₂ Molecular Weight: 170,1297 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077609 CAS-Nummer: 371-27-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...