Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate 97%

BLDPBD634642-1GEA 62 EUR
BLDPBD634642-1G BLDPBD634642-100MG BLDPBD634642-5G BLDPBD634642-250MG
Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate 97%
Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₁₂N₂O₄
Molecular Weight: 272,26 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11042478
CAS-Nummer: 142593-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR