2-(Phenylthio)thiophen 95%
Lieferant: BLD PHARMATECH GMBH
BLDPBD63459-100MGEA
19
EUR
BLDPBD63459-100MG
BLDPBD63459-250MG
2-(Phenylthio)thiophen 95%
2-(Phenylthio)thiophene
Formel:
C₁₀H₈S₂ Molecular Weight: 192,30 g/mol |
MDL:
MFCD00052141 CAS-Nummer: 16718-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...