(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) 98% 99%ee

BLDPBD634124-50MGEA 19 EUR
BLDPBD634124-50MG BLDPBD634124-100MG BLDPBD634124-1G BLDPBD634124-250MG
(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) 98% 99%ee
(R,R)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline)

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₅H₂₆N₂O₂
Molecular Weight: 266,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28038687
CAS-Nummer: 150529-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR