3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine 97%

BLDPBD633329-250MGEA 29 EUR
BLDPBD633329-250MG BLDPBD633329-1G BLDPBD633329-5G
3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine 97%
3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₂O₂S
Molecular Weight: 184,25 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08705268
CAS-Nummer: 255901-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR