UNC0224 98%

BLDPBD629978-25MGEA 130 EUR
BLDPBD629978-25MG BLDPBD629978-100MG BLDPBD629978-50MG
UNC0224 98%
UNC0224

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₆H₄₃N₇O₂
Molecular Weight: 485,67 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18251564
CAS-Nummer: 1197196-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR