2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene] 97%

BLDPBD629315-250MGEA 8 EUR
BLDPBD629315-250MG BLDPBD629315-1G BLDPBD629315-0.025 BLDPBD629315-25G BLDPBD629315-10G BLDPBD629315-5G
2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene] 97%
2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene]

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₅H₁₅BrO
Molecular Weight: 411,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29050113
CAS-Nummer: 899422-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR