N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine 98%

BLDPBD627873-250MGEA 29 EUR
BLDPBD627873-250MG BLDPBD627873-1G BLDPBD627873-100MG BLDPBD627873-5G
N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine 98%
N,N',N''-Tri(m-tolyl)-1,3,5-triazine-2,4,6-triamine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₂₄H₂₄N₆
Molecular Weight: 396,50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19440763
CAS-Nummer: 82504-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR