2-(6-Hydroxybenzofuran-3-yl)acetic acid 95%

BLDPBD62747-25GEA 84 EUR
BLDPBD62747-25G BLDPBD62747-5G BLDPBD62747-10G BLDPBD62747-1G BLDPBD62747-100G
2-(6-Hydroxybenzofuran-3-yl)acetic acid 95%
2-(6-Hydroxybenzofuran-3-yl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₈O₄
Molecular Weight: 192,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05273635
CAS-Nummer: 69716-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR