3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole 98%

BLDPBD625749-100MGEA 46 EUR
BLDPBD625749-100MG
3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole 98%
3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₄H₁₅Br₂N
Molecular Weight: 477,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22571691
CAS-Nummer: 1345021-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR