3-Bromo-9,9-diphenyl-9H-fluorene 98%

BLDPBD625737-1GEA 23 EUR
BLDPBD625737-1G BLDPBD625737-25G BLDPBD625737-250MG BLDPBD625737-5G
3-Bromo-9,9-diphenyl-9H-fluorene 98%
3-Bromo-9,9-diphenyl-9H-fluorene

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₅H₁₇Br
Molecular Weight: 397,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24849694
CAS-Nummer: 1547491-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR