4-(Methylsulfonyl)-2-nitrobenzoesäure 98%
|
|
BLDPBD6246-500GEA
150
EUR
BLDPBD6246-500G
BLDPBD6246-1000G
BLDPBD6246-25G
BLDPBD6246-10G
BLDPBD6246-100G
BLDPBD6246-0.025G
BLDPBD6246-5G
4-(Methylsulfonyl)-2-nitrobenzoesäure 98%
4-(Methylsulphonyl)-2-nitrobenzoic acid
Storage condition: Sealed in dry, Room Temperature
Formel:
C₈H₇NO₆S Molecular Weight: 245,2100 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01941299 CAS-Nummer: 110964-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...