N-Methyl-2-nitro-4-(trifluormethyl)anilin 98%

BLDPBD6135-5GEA 18 EUR
BLDPBD6135-5G
N-Methyl-2-nitro-4-(trifluormethyl)anilin 98%
N-Methyl-2-nitro-4-(trifluormethyl)anilin
Formel: C₈H₇F₃N₂O₂
Molecular Weight: 220,1486 g/mol
MDL: MFCD00798375
CAS-Nummer: 20200-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR