5-Brom-o-toluylsäure 98%

BLDPBD6042-100GEA 59 EUR
BLDPBD6042-100G BLDPBD6042-25G BLDPBD6042-5G BLDPBD6042-1G BLDPBD6042-500G BLDPBD6042-10G
5-Brom-o-toluylsäure 98%
5-Brom-o-toluylsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00267350
CAS-Nummer: 79669-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR