1-Phenyl-3-buten-1-ol 98%

BLDPBD60170-1GEA 69 EUR
BLDPBD60170-1G BLDPBD60170-5G BLDPBD60170-25G
1-Phenyl-3-buten-1-ol 98%
4-Phenyl-1-buten-4-ol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₂O
Molecular Weight: 148,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039617
CAS-Nummer: 936-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR