1-Pentansulfonsäure Natriumsalz 98%

Synonyme: Natriumpentylsulfonat

BLDPBD60035-100GEA 87 EUR
BLDPBD60035-100G BLDPBD60035-10G BLDPBD60035-25G BLDPBD60035-5G BLDPBD60035-500G BLDPBD60035-0.5G
1-Pentansulfonsäure Natriumsalz 98%
1-Pentanesulphonic acid sodium salt

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₁O₃SNa
Molecular Weight: 174,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007541
CAS-Nummer: 22767-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR