CZC-25146 98%

BLDPBD600050-250MGEA 316 EUR
BLDPBD600050-250MG BLDPBD600050-50MG BLDPBD600050-1G BLDPBD600050-100MG
CZC-25146 98%
CZC-25146

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂₂H₂₅FN₆O₄S
Molecular Weight: 488,54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28160475
CAS-Nummer: 1191911-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR