1H,1H,2H,2H-Perfluoroctyltriethoxysilan 96%

BLDPBD58475-25GEA 63 EUR
BLDPBD58475-25G BLDPBD58475-500G BLDPBD58475-1G BLDPBD58475-100G BLDPBD58475-5G BLDPBD58475-10G
1H,1H,2H,2H-Perfluoroctyltriethoxysilan 96%
1H,1H,2H,2H-Perfluoroctyltriethoxysilan

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₄H₁₉F₁₃O₃Si
Molecular Weight: 510,367 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042333
CAS-Nummer: 51851-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR