4-Chloro-N-methylbenzamide 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD583201-1GEA 206 EUR
BLDPBD583201-1G BLDPBD583201-5G
4-Chloro-N-methylbenzamide 98%
4-Chloro-N-methylbenzamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₈ClNO
Molecular Weight: 169,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00774485
CAS-Nummer: 6873-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR