5-(Trifluormethoxy)salicylaldehyd 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD5758-10GEA 93 EUR
BLDPBD5758-10G BLDPBD5758-1G BLDPBD5758-5G BLDPBD5758-25G
5-(Trifluormethoxy)salicylaldehyd 95%
5-(Trifluormethoxy)salicylaldehyd

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₅F₃O₃
Molecular Weight: 206,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075249
CAS-Nummer: 93249-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR