5-Bromisophthalsäure 97%

Synonyme: 5-Brom-1,3-benzoldicarbonsäure

BLDPBD5697-100GEA 278 EUR
BLDPBD5697-100G BLDPBD5697-5G BLDPBD5697-1G BLDPBD5697-10G BLDPBD5697-500G BLDPBD5697-25G
5-Bromisophthalsäure 97%
5-Bromisophthalsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₅BrO₄
Molecular Weight: 245,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00152019
CAS-Nummer: 23351-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR