5-Bromopyrido[3,4-b]pyrazine 95%

BLDPBD562040-1GEA 745 EUR
BLDPBD562040-1G BLDPBD562040-100MG BLDPBD562040-250MG
5-Bromopyrido[3,4-b]pyrazine 95%
5-Bromopyrido[3,4-b]pyrazine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₄BrN₃
Molecular Weight: 210,031 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12401005
CAS-Nummer: 1159829-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR