11H-Benzo[a]carbazole 98%

BLDPBD55823-5GEA 105 EUR
BLDPBD55823-5G BLDPBD55823-25G BLDPBD55823-1G
11H-Benzo[a]carbazole 98%
11H-Benzo[a]carbazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₆H₁₁N
Molecular Weight: 217,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215939
CAS-Nummer: 239-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR