Chloro-IB-MECA 99+%

BLDPBD55443-50MGEA 530 EUR
BLDPBD55443-50MG
Chloro-IB-MECA 99+%
Chloro-IB-MECA

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₈H₁₈ClIN₆O₄
Molecular Weight: 544,73 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01861178
CAS-Nummer: 163042-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR