Methyl 4,4,4-Trichlorobutanoate 97%

BLDPBD553993-1GEA 126 EUR
BLDPBD553993-1G BLDPBD553993-250MG BLDPBD553993-5G
Methyl 4,4,4-Trichlorobutanoate 97%
Methyl 4,4,4-Trichlorobutanoate

Storage condition: Sealed in dry, 2-8°C

Formel: C₅H₇Cl₃O₂
Molecular Weight: 205,467 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28144036
CAS-Nummer: 19376-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR