(R)-(+)-1-Boc-3-aminopyrrolidin 98%

BLDPBD5526-5GEA 25 EUR
BLDPBD5526-5G BLDPBD5526-25G BLDPBD5526-10G BLDPBD5526-1G
(R)-(+)-1-Boc-3-aminopyrrolidin 98%
(R)-(+)-1-Boc-3-aminopyrrolidin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₈N₂O₂
Molecular Weight: 186,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419272
CAS-Nummer: 147081-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR