1,4,5,8-Tetrachloroanthraquinone 95%

BLDPBD54340-25GEA 344 EUR
BLDPBD54340-25G BLDPBD54340-5G BLDPBD54340-1G
1,4,5,8-Tetrachloroanthraquinone 95%
1,4,5,8-Tetrachloroanthraquinone

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₄Cl₄O₂
Molecular Weight: 345,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035801
CAS-Nummer: 81-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR